Skip to Content
Merck
All Photos(1)

Key Documents

570362

Sigma-Aldrich

2-(9H-Carbazol-9-yl)ethyl acrylate

97%

Synonym(s):

2-(9H-Carbazol-9-yl)ethyl methacrylate, 9H-Carbazole-9-ethylacrylate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$542.00

$542.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$542.00

About This Item

Empirical Formula (Hill Notation):
C17H15NO2
CAS Number:
Molecular Weight:
265.31
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

$542.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

mp

77-81 °C (lit.)

storage temp.

2-8°C

SMILES string

C=CC(=O)OCCn1c2ccccc2c3ccccc13

InChI

1S/C17H15NO2/c1-2-17(19)20-12-11-18-15-9-5-3-7-13(15)14-8-4-6-10-16(14)18/h2-10H,1,11-12H2

InChI key

KOHMQTTZAWPDNE-UHFFFAOYSA-N

General description

2-(9H-Carbazol-9-yl)ethyl acrylate (PCz) is an acrylate based conducting polymer with carbazole as electron donating pendant group. It can be used as a charge transporting material as it exhibits high charge carrier mobility and photochemical stability. It also has high thermal and electroluminescent properties that make it useful in organic electronics based application.[1][2][3]

Application

PCz can be used as a conducting polymer in the fabrication of a variety of devices which include organic light emitting diodes (OLEDs), photovoltaic cells and memory based devices.[1][4][5]

Features and Benefits

NLO chromophore

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reversible addition-fragmentation chain transfer polymerization of methacrylates containing hole-or electron-transporting groups
Zhao P, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 45(2), 242-252 (2007)
Synthesis and switching properties of photochromic carbazole-spironaphthoxazine copolymer
Wang S, et al.
Journal of Photochemistry and Photobiology A: Chemistry, 192(1), 17-22 (2007)
Single component transparent free-standing films based on polyhedral octasilicate-core dendrimers bearing carbazole terminal groups and their emission properties
Irie Y and Naka K
Journal of Polymer Science Part A: Polymer Chemistry, 54(5), 628-633 (2016)
Non-volatile WORM memory device based on an acrylate polymer with electron donating carbazole pendant groups
Teo EYH, et al.
Organic Electronics, 7(3), 173-180 (2006)
Poly(methyl methacrylate)copolymers containing pendant carbazole and oxadiazole moieties for applications in single-layer organic light emitting devices.
Evanoff DD, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 46(23), 7882-7897 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service