Skip to Content
Merck
All Photos(1)

Key Documents

765120

Sigma-Aldrich

1-Pyrenemethyl methacrylate

contains ≤200 ppm MEHQ as inhibitor, 99% (GC)

Synonym(s):

1-Pyrenylmethyl methacrylate, Pyrene methacrylate, Pyrenylmethyl methacrylate

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$207.00

$207.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$207.00

About This Item

Empirical Formula (Hill Notation):
C21H16O2
CAS Number:
Molecular Weight:
300.35
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$207.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

99% (GC)

form

powder

contains

≤200 ppm MEHQ as inhibitor

mp

100-105 °C

storage temp.

2-8°C

SMILES string

CC(=C)C(=O)OCc1ccc2ccc3cccc4ccc1c2c34

InChI

1S/C21H16O2/c1-13(2)21(22)23-12-17-9-8-16-7-6-14-4-3-5-15-10-11-18(17)20(16)19(14)15/h3-11H,1,12H2,2H3

InChI key

BUQDPCFXOBQDLX-UHFFFAOYSA-N

General description

1-Pyrenemethyl methacrylate is a fluorescent methacrylate monomer which is used for synthesising poly methyl methacrylate (PMMA) based optical and fluorescent temperature sensors. These polymeric sensors exhibit reverse solubility transition wherein the polymer is insoluble at low temperatures and is soluble at higher temperatures. These sensors are used as biosensors, in drug delivery, logical gates and optical sensing applications.[1]

Application

  • fluorescent monomer

Features and Benefits

Water-soluble, thermo-responsive and responds to minor local environment changes.[1]

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

PMMA based soluble polymeric temperature sensors based on UCST transition and solvatochromic dyes
Pietsch, C., Hoogenboom, R., & Schubert, U. S.
Polym. Chem., 1(7), 1005-1008 (2010)
A fluorescent thermometer based on a pyrene-labeled thermoresponsive polymer
Pietsch, C., Vollrath, A., Hoogenboom, R., & Schubert, U. S.
Sensors, 10(9), 7979-7990 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service