Skip to Content
Merck
All Photos(1)

Key Documents

56776

Sigma-Aldrich

Trihexyltetradecylphosphonium dicyanamide

≥95% (qNMR)

Synonym(s):

Tetradecyltrihexylphosphonium dicyanamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[CH3(CH2)5]3P[N(CN)2](CH2)13CH3
CAS Number:
Molecular Weight:
549.90
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95% (qNMR)

density

0.90 g/mL at 20 °C (lit.)

functional group

amine
phosphine

SMILES string

N#C[N-]C#N.CCCCCCCCCCCCCC[P+](CCCCCC)(CCCCCC)CCCCCC

InChI

1S/C32H68P.C2N3/c1-5-9-13-17-18-19-20-21-22-23-24-28-32-33(29-25-14-10-6-2,30-26-15-11-7-3)31-27-16-12-8-4;3-1-5-2-4/h5-32H2,1-4H3;/q+1;-1

InChI key

DOMOOBQQQGXLGU-UHFFFAOYSA-N

Application

Trihexyltetradecylphosphonium dicyanamide is an ionic liquid that can be used:
  • As an extraction solvent for the separation of organic compounds.
  • In the experiment to study the solubility of carbon dioxide and binary gas mixtures in ionic liquids.
  • To prepare hybrid ionogels having electrochemical applications.
  • As a solvent for the extraction of aromatics from pyrolytic sugars.

Caution

may become slightly turbid on storage

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Application of trihexyltetradecylphosphonium dicyanamide ionic liquid for various types of separations problems: Activity coefficients at infinite dilution measurements utilizing GLC method
Kabane B and Redhi GG
Fluid Phase Equilibria, 493, 181-187 (2019)
Solubility of CO2/CH4 gas mixtures in ionic liquids
Ramdin M, et al.
Fluid Phase Equilibria, 375, 134-142 (2014)
High pressure solubility of CO2 in non-fluorinated phosphonium-based ionic liquids
Ramdin M, et al.
Journal of Supercritical Fluids, 82, 41-49 (2013)
Photo-patternable hybrid ionogels for electrochromic applications
Kavanagh A, et al.
Journal of Materials Chemistry, 21(24), 8687-8693 (2011)
Xiaohua Li et al.
Bioresource technology, 216, 12-18 (2016-05-24)
Fermentative bioethanol production from pyrolytic sugars was improved via aromatics removal by liquid-liquid extraction. As solvents, the ionic liquid (IL) trihexyltetradecylphosphonium dicyanamide (P666,14[N(CN)2]) and ethyl acetate (EA) were compared. Two pyrolytic sugar solutions were created from acid-leached and untreated pinewood

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service