567620
2-Chloro-4-(trifluoromethyl)benzonitrile
97%
Synonym(s):
3-Chloro-4-cyanobenzotrifluoride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
ClC6H3(CF3)CN
CAS Number:
Molecular Weight:
205.56
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
refractive index
n20/D 1.4840 (lit.)
bp
192-193 °C (lit.)
density
1.389 g/mL at 25 °C (lit.)
SMILES string
FC(F)(F)c1ccc(C#N)c(Cl)c1
InChI
1S/C8H3ClF3N/c9-7-3-6(8(10,11)12)2-1-5(7)4-13/h1-3H
InChI key
GEHMLBFNZKJDQM-UHFFFAOYSA-N
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
219.9 °F - closed cup
Flash Point(C)
104.4 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service