Skip to Content
Merck
All Photos(1)

Key Documents

542636

Sigma-Aldrich

Methyl 2,6-difluorobenzoate

97%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$92.90

$92.90


Estimated to ship onMay 25, 2025


Request a Bulk Order

Select a Size

Change View
25 G
$92.90

About This Item

Linear Formula:
F2C6H3CO2CH3
CAS Number:
Molecular Weight:
172.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$92.90


Estimated to ship onMay 25, 2025


Request a Bulk Order

Assay

97%

refractive index

n20/D 1.476 (lit.)

bp

203-204 °C (lit.)

density

1.281 g/mL at 25 °C (lit.)

functional group

ester
fluoro

SMILES string

COC(=O)c1c(F)cccc1F

InChI

1S/C8H6F2O2/c1-12-8(11)7-5(9)3-2-4-6(7)10/h2-4H,1H3

InChI key

QNPFLTKQLFSKBY-UHFFFAOYSA-N

General description

Methyl 2,6-difluorobenzoate is a fluorinated aromatic ester. It can be synthesized from 2,6-difluorobenzoic acid via esterification with methanol.[1]

Application

Methyl 2,6-difluorobenzoate may be used in the synthesis of 9-hydroxy-13-(2,6-difluorophenyl)-3H-dibenzo[b,i]xanthen-3-one[2] and 2,6-difluorobenzohydroxamic acid.[2]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

203.0 °F - closed cup

Flash Point(C)

95 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Conformational analysis. Part 25. The evaluation of molecular geometries by the lanthanide induced shift (LIS) technique.
Abraham RJ, et al.
J. Chem. Soc. Perkin Trans. II, 11, 1973-1979 (1995)
2, 6-Difluorobenzohydroxamic acid.
Meng XG and Shang XM.
Acta Crystallographica Section B, Structural Science, Crystal Engineering and Materials, 62(2), o651-o652 (2006)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service