Skip to Content
Merck
All Photos(1)

Key Documents

560855

Sigma-Aldrich

10-Bromo-1-decene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br(CH2)8CH=CH2
CAS Number:
Molecular Weight:
219.16
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
97%

Quality Level

Assay

97%

refractive index

n20/D 1.4660 (lit.)

bp

50 °C/0.3 mmHg (lit.)

density

1.092 g/mL at 25 °C (lit.)

functional group

alkyl halide
allyl
bromo

SMILES string

BrCCCCCCCCC=C

InChI

1S/C10H19Br/c1-2-3-4-5-6-7-8-9-10-11/h2H,1,3-10H2

InChI key

JVVPJOMYWVYPOF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

10-Bromo-1-decene can be synthesized by reacting 9-decen-1-ol with PBr3. The product undergoes reduction reaction with 2-propylbenzo[d][1,3,2]dioxaborole (PBD) and Bu3SnH under room temperature conditions to yield 1-decene.

Packaging

10-Bromo-1-decene may be used to synthesize:
  • 4′-(9-decenyloxy) biphenyl-4-carboxylic acid methyl ester
  • 4′-(9-decenyloxy)biphenyl-4-carboxylic acid
  • 9-decenylmagnesium bromide, which was employed for the preparation of (R)-tridec-12-en-2-ol

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

206.0 °F - closed cup

Flash Point(C)

96.67 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julian Gebauer et al.
The Journal of organic chemistry, 71(5), 2021-2025 (2006-02-25)
A simple access to gamma,delta-unsaturated-beta-keto lactones is presented, allowing a rapid total synthesis of the naturally occurring 16-membered macrolide antibiotic (-)-A26771B via cross metathesis, asymmetric dihydroxylation, and lactonization as the key steps.
The four-membered-ring chemical shift anomaly.
Lambert JB, et al.
The Journal of Organic Chemistry, 48(22), 3982-3985 (1983)
Room temperature ferroelectric terpolymers with large spontaneous polarization.
Naciri J, et al.
Macromolecules, 28(15), 5274-5279 (1995)
Novel ferroelectric and electroclinic organosiloxane liquid crystals.
Naciri J, et al.
Chemistry of Materials, 7(7), 1397-1402 (1995)
Radical initiation using borole derivatives.
Montgomery I, et al.
Tetrahedron Letters, 49(4), 628-630 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service