Skip to Content
Merck
All Photos(2)

Key Documents

559954

Sigma-Aldrich

2,6-Difluorobenzyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
F2C6H3CH2Cl
CAS Number:
Molecular Weight:
162.56
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

34-38 °C (lit.)

functional group

chloro
fluoro

SMILES string

Fc1cccc(F)c1CCl

InChI

1S/C7H5ClF2/c8-4-5-6(9)2-1-3-7(5)10/h1-3H,4H2

InChI key

MJXRENZUAQXZGJ-UHFFFAOYSA-N

General description

Gaseous chlorine reacts with 2,6-difluorotoluene to yield 2,6-difluorobenzyl chloride.[1]

Application

2,6-Difluorobenzyl chloride may be used to synthesize 2,6-difluorobenzaldehyde[1] and enantiopure methiozolins.[2]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Preparation of 2, 6-difluoro-n-alkylbenzenes from 1, 3-difluorobenzene Transformation of 2, 6-difluorotoluene to the corresponding benzaldehyde via benzyl chloride.
Malykhin EV and Shteingarts VD.
Journal of Fluorine Chemistry, 91(1), 19-20 (1998)
Synthesis and herbicidal activities of enantiopure methiozolins.
Nam JH, et al.
Bull. Korean Chem. Soc., 33, 297-300 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service