Skip to Content
Merck
All Photos(1)

Key Documents

557331

Sigma-Aldrich

3-Ethynyl-α,α,α-trifluorotoluene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HC≡CC6H4CF3
CAS Number:
Molecular Weight:
170.13
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.464 (lit.)

bp

146 °C (lit.)

density

1.178 g/mL at 25 °C (lit.)

functional group

fluoro

SMILES string

FC(F)(F)c1cccc(c1)C#C

InChI

1S/C9H5F3/c1-2-7-4-3-5-8(6-7)9(10,11)12/h1,3-6H

InChI key

PAHXLHWOHJTWRU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Ethynyl-α,α,α-trifluorotoluene may be used to synthesize:
  • 1-thioacetyl-4[3-trifluoromethyl-1-(ethynyl)phenyl]benzene[1]
  • 4-{[4-(3-(trifluoromethyl)phenyl)-1H-1,2,3-triazol-1-yl]methyl}benzoic acid[2]
  • 4-{4-[3-(trifluoromethyl)phenyl]-1H-1,2,3-triazol-1-yl}benzoic acid[2]

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

69.8 °F - closed cup

Flash Point(C)

21 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis, structural analysis, and self-assembly of phenylene ethynylene oligomers and their F, CF3, and CH3 substituted derivatives.
Percec S, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 42(3), 541-550 (2004)
Matthias Wrobel et al.
Beilstein journal of organic chemistry, 8, 1027-1036 (2012-09-29)
A series of substituted diaryltriazoles was prepared by a solid-phase-synthesis protocol using a modified Wang resin. The copper(I)- or ruthenium(II)-catalyzed 1,3-cycloaddition on the polymer bead allowed a rapid synthesis of the target compounds in a parallel fashion with in many

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service