Skip to Content
Merck
All Photos(1)

Documents

553425

Sigma-Aldrich

1-Bromo-4-(difluoromethoxy)benzene

95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4BrOCHF2
CAS Number:
Molecular Weight:
223.01
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

refractive index

n20/D 1.502 (lit.)

density

1.631 g/mL at 25 °C (lit.)

SMILES string

FC(F)Oc1ccc(Br)cc1

InChI

1S/C7H5BrF2O/c8-5-1-3-6(4-2-5)11-7(9)10/h1-4,7H

InChI key

ORIYZUFTROJBQJ-UHFFFAOYSA-N

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

93.0 °F - closed cup

Flash Point(C)

33.9 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service