Skip to Content
Merck
All Photos(2)

Key Documents

551260

Sigma-Aldrich

5-Oxo-1-phenyl-2-pyrazolin-3-carboxylic acid

Synonym(s):

1-Phenyl-3-carboxy-5-pyrazolone, 3-Carboxy-1-phenyl-2-pyrazolin-5-one

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$233.00

$233.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$233.00

About This Item

Empirical Formula (Hill Notation):
C10H8N2O3
CAS Number:
Molecular Weight:
204.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$233.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

form

solid

mp

237 °C (lit.)

functional group

carboxylic acid

SMILES string

OC(=O)C1=NN(C(=O)C1)c2ccccc2

InChI

1S/C10H8N2O3/c13-9-6-8(10(14)15)11-12(9)7-4-2-1-3-5-7/h1-5H,6H2,(H,14,15)

InChI key

IMZSHPUSPMOODC-UHFFFAOYSA-N

Application

Reactant involved in synthesis of:
  • Pyrazolone derivatives for antiprion activity via heterocyclocondensation
  • Heterocyclic thiazolidinone and azetidinone compound for antibacterial studies

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service