Skip to Content
Merck
All Photos(2)

Key Documents

451436

Sigma-Aldrich

3-Mercaptobenzoic acid

95%

Synonym(s):

3-Sulfanylbenzoic acid, m-Mercaptobenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSC6H4CO2H
CAS Number:
Molecular Weight:
154.19
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

95%

form

solid

mp

144-147 °C (lit.)

functional group

carboxylic acid

SMILES string

OC(=O)c1cccc(S)c1

InChI

1S/C7H6O2S/c8-7(9)5-2-1-3-6(10)4-5/h1-4,10H,(H,8,9)

InChI key

RSFDFESMVAIVKO-UHFFFAOYSA-N

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leonora Velleman et al.
Physical chemistry chemical physics : PCCP, 13(43), 19587-19593 (2011-10-11)
Electroless deposition was used to coat porous alumina membranes with gold. This process reduced the pore diameters and provided a platform suitable for surface modifications with self assembled monolayers (SAMs). The surface enhanced Raman scattering (SERS) effect was employed in
A catalyst-free system for 3-sulfenylation of free (N-H) indoles with mercaptobenzoic acid under alkaline conditions.
Zhang X, et al.
Royal Society of Chemistry Advances, 3(44), 22280-22284 (2013)
P S Robbins et al.
Die Naturwissenschaften, 90(11), 517-520 (2003-11-12)
The female-produced sex pheromone of Phyllophaga crinita (Burmeister) (Coleoptera: Scarabaeidae: Melolonthinae; the adult has no common name) is identified as methyl 2-(methylthio)benzoate. This is the first identification of a sulfur-containing, long-distance, female-produced sex attractant from any insect taxa. The root-feeding
Homan Kang et al.
Scientific reports, 5, 10144-10144 (2015-05-29)
Recently, preparation and screening of compound libraries remain one of the most challenging tasks in drug discovery, biomarker detection, and biomolecular profiling processes. So far, several distinct encoding/decoding methods such as chemical encoding, graphical encoding, and optical encoding have been
Isabel Freitag et al.
Journal of biomedical optics, 20(5), 55002-55002 (2015-05-06)
Identification of tumor and normal cells is a promising application of Raman spectroscopy. The throughput of Raman-assisted cell sorting is limited by low sensitivity. Surface-enhanced Raman spectroscopy (SERS) is a well-recognized candidate to increase the intensity of Raman signals of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service