Skip to Content
Merck
All Photos(2)

Documents

551112

Sigma-Aldrich

Rubrene

sublimed grade, 99.99% trace metals basis

Synonym(s):

5,6,11,12-Tetraphenylnaphthacene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H28
CAS Number:
Molecular Weight:
532.67
Beilstein:
1917339
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

grade

sublimed grade

Assay

99.99% trace metals basis

mp

330-335 °C (lit.)

λmax

299 nm
299 nm

fluorescence

λem 550 nm in THF

Orbital energy

HOMO 5.4 eV 
LUMO 3.2 eV 

OLED Device Performance

ITO/CuPc/NPD/Alq3:Rubrene (5%): DCM2 (2%)/Alq3/Mg:In

  • Color: red
  • Max. Luminance: 7780 Cd/m2

ITO/PEDOT:PSS/EHCz:Rubrene (1 wt%)/Cs2CO3:ITO
  • Color: red
  • Max. EQE: 0.03 %

SMILES string

c1ccc(cc1)-c2c3ccccc3c(-c4ccccc4)c5c(-c6ccccc6)c7ccccc7c(-c8ccccc8)c25

InChI

1S/C42H28/c1-5-17-29(18-6-1)37-33-25-13-14-26-34(33)39(31-21-9-3-10-22-31)42-40(32-23-11-4-12-24-32)36-28-16-15-27-35(36)38(41(37)42)30-19-7-2-8-20-30/h1-28H

InChI key

YYMBJDOZVAITBP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

TGA/DSC Lot specific traces available upon request

Application

Organic electronic material useful as OLED dopant (red, λem = 550nm) and as p-type organic semiconductor. Carrier mobilities of 8-20 cm2/ Vs can be achieved in OFETs based on single crystals of sublimed rubrene.
Reagent for chemiluminescence research and for transition metal complex ligation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Morpurgo, A. F. et al.
Physica Status Solidi (A): Applied Research, 201, 1302-1302 (2004)
N R Armstrong et al.
Annual review of physical chemistry, 52, 391-422 (2001-04-28)
Electrochemical processes leading to light emission are reviewed, with emphasis on aspects of this subject relevant to the understanding and optimization of electrogenerated luminescence (EL) in organic thin-film materials. The basic energetic requirements of light emission from electrochemically initiated solution
Alexander S Bieber et al.
The Journal of chemical physics, 153(8), 084703-084703 (2020-09-03)
Triplet-triplet annihilation-based photon upconversion (UC) using bulk perovskite sensitizers has been previously shown to facilitate efficient UC at low fluences. However, the fabrication of the UC devices has not been fully optimized; thus, there is room for improvement. Here, we
V Podzorov et al.
Physical review letters, 95(22), 226601-226601 (2005-12-31)
We have observed the Hall effect in the field-induced accumulation layer on the surface of single-crystal samples of a small-molecule organic semiconductor rubrene. The Hall mobility muH increases with decreasing temperature in both the intrinsic (high-temperature) and trap-dominated (low-temperature) conduction
Yuanzhen Chen et al.
Advanced materials (Deerfield Beach, Fla.), 23(45), 5370-5375 (2011-10-18)
Commonly observed variations in photoluminescence (PL) spectra of crystalline organic semiconductors, including the appearance or enhancement of certain PL bands, are shown to originate from a small amount of structural disorder (e.g., amorphous inclusions embedded in a crystal), rather than

Articles

Sublimed materials for organic electronic devices such of OFETs and OTFTs allow the achievement of better electronic properties, and may help increase a device’s lifetime.

In this article, we demonstrate that bis-styrylbenzene derivatives show promising characteristics for very low lasing thresholds and discuss the design considerations for organic lasing molecules.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service