Skip to Content
Merck
All Photos(5)

Key Documents

264199

Sigma-Aldrich

9,10-Bis(phenylethynyl)anthracene

greener alternative

97%

Synonym(s):

BPEA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H18
CAS Number:
Molecular Weight:
378.46
Beilstein:
1891432
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Pricing and availability is not currently available.

Assay

97%

greener alternative product score

old score: 37
new score: 13
Find out more about DOZN™ Scoring

greener alternative product characteristics

Design for Energy Efficiency
Catalysis
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

248-250 °C (lit.)

greener alternative category

SMILES string

c1ccc(cc1)C#Cc2c3ccccc3c(C#Cc4ccccc4)c5ccccc25

InChI

1S/C30H18/c1-3-11-23(12-4-1)19-21-29-25-15-7-9-17-27(25)30(28-18-10-8-16-26(28)29)22-20-24-13-5-2-6-14-24/h1-18H

InChI key

ZHBOFZNNPZNWGB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product belongs to category of Re-engineered products, showing key improvements in Green Chemistry Principles “Design for Energy Efficiency” and “Enhanced Catalysis”. Click here to view its DOZN scorecard.

Application

Reagent for chemiluminescence research.[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sanjib Jana et al.
Physical chemistry chemical physics : PCCP, 21(26), 14341-14349 (2019-02-01)
Polyacenes, such as anthracene, tetracene, pentacene etc., have been identified as potential candidates for singlet fission (SF) and triplet-triplet annihilation (TTA) processes in their crystalline and thin film forms as they possess significant singlet and triplet exciton couplings. Interestingly, phenyl-ethynyl
Massimo Di Fusco et al.
Analytical and bioanalytical chemistry, 407(6), 1567-1576 (2014-12-30)
Doped organically modified silica nanoparticles (ORMOSIL NPs) with luminescent molecules represent a potent approach to signal amplification in biomolecule labeling. Herein, we report the synthesis of new ORMOSIL NPs incorporating thermochemiluminescent (TCL) 1,2-dioxetane derivatives to prepare TCL labels for ultrasensitive
Sukumaran Santhosh Babu et al.
Nature communications, 4, 1969-1969 (2013-06-06)
Nonvolatile room-temperature luminescent molecular liquids are a new generation of organic soft materials. They possess high stability, versatile optical properties, solvent-free fluid behaviour and can effectively accommodate dopant dye molecules. Here we introduce an approach to optimize anthracene-based liquid materials
Dong Yang et al.
Nature communications, 8, 15727-15727 (2017-06-07)
Transfer of both chirality and energy information plays an important role in biological systems. Here we show a chiral donor π-gelator and assembled it with an achiral π-acceptor to see how chirality and energy can be transferred in a composite
Si-Hui Zhuang et al.
Journal of fluorescence, 26(1), 317-322 (2015-11-14)
The unique photoproperties of quantum dots are promising for potential application in bioassays. In the present study, quantum dots were applied to a luminescent oxygen channeling assay. The reaction system developed in this study was based on interaction of biotin

Articles

Graphene is the building block for carbon nanomaterials with different dimensionalities.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service