Skip to Content
Merck
All Photos(1)

Documents

549312

Sigma-Aldrich

3,3′-Dihydroxydiphenyl disulfide

97%

Synonym(s):

Bis(3-hydroxyphenyl) disulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
S2(C6H4OH)2
CAS Number:
Molecular Weight:
250.34
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

89-93 °C (lit.)

SMILES string

Oc1cccc(SSc2cccc(O)c2)c1

InChI

1S/C12H10O2S2/c13-9-3-1-5-11(7-9)15-16-12-6-2-4-10(14)8-12/h1-8,13-14H

InChI key

XBNOMKROXZGMFW-UHFFFAOYSA-N

General description

3,3′-Dihydroxydiphenyl disulfide [bis(3-hydroxyphenyl) disulfide] is a symmetrical disulfide derivative that can be prepared by treating 3-hydroxythiophenol with ferric chloride.

Application

3,3′-Dihydroxydiphenyl disulfide may be used in the preparation of bis-[3-(4-fluorobenzyloxy)-phenyl]-disulfide by reacting with 4-fluorobenzyl bromide.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polar Conjugative Effect of Aryldithia Group on the Dissociation and Ultraviolet Spectra of Substituted Phenols.
Oae S and Yoshihara M.
Bulletin of the Chemical Society of Japan, 41(9), 2082-2086 (1968)
Efficient Route for the Preparation of Benzyloxy-Substituted Benzenesulfonyl Chlorides from Mercaptophenols.
Jumeaux C, et al.
Synthetic Communications, 41(12), 1805-1808 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service