Skip to Content
Merck
All Photos(3)

Key Documents

160326

Sigma-Aldrich

4,4′-Bis(diethylamino)benzophenone

≥99%

Synonym(s):

4,4′-(Tetraethyldiamino)benzophenone, 4,4′-Bis(diethylamino)diphenyl ketone, p ,p ′-Tetraethyldiaminobenzophenone, Bis[4-(diethylamino)phenyl]methanone, Michler′s ethyl ketone

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$317.00
100 G
$512.00

$317.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$317.00
100 G
$512.00

About This Item

Linear Formula:
[(C2H5)2NC6H4]2CO
CAS Number:
Molecular Weight:
324.46
Beilstein:
2148611
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

$317.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

≥99%

mp

89-92 °C (lit.)

SMILES string

CCN(C1=CC=C(C(C2=CC=C(N(CC)CC)C=C2)=O)C=C1)CC

InChI

1S/C21H28N2O/c1-5-22(6-2)19-13-9-17(10-14-19)21(24)18-11-15-20(16-12-18)23(7-3)8-4/h9-16H,5-8H2,1-4H3

InChI key

VYHBFRJRBHMIQZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • The effect of 4, 4′-bis (N, N-diethylamino) benzophenone on the degree of conversion in liquid photopolymer for dental 3D printing: Examines its use as a co-initiator in dental 3D printing materials (DH Lee et al., 2015).
  • Ketone Number and Substitution Effect of Benzophenone Derivatives on the Free Radical Photopolymerization: Compares its performance with other photoinitiators in polymerization (TL Huang, YC Chen, 2021).
  • Michler′s ethylketone as a novel negative-ion matrix for the enhancement of lipid MALDI tissue imaging: Uses 4,4′-bis(diethylamino)benzophenone in matrix-assisted laser desorption/ionization (MALDI) for tissue imaging (Y Shi et al., 2022).

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Thi Kim Hoang Trinh et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 25(39), 9242-9252 (2019-04-26)
In the search of smarter routes to control the conditions of N-heterocyclic carbene (NHCs) formation, a two-component air-stable NHC photogenerating system is reported. It relies on the irradiation at 365 nm of a mixture of 2-isopropylthioxanthone (ITX) with 1,3-bis(mesityl)imidazoli(ni)um tetraphenylborate. The

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service