Skip to Content
Merck
All Photos(1)

Key Documents

548898

Sigma-Aldrich

2,3-Dihydro-3-oxo-4H-1,4-benzoxazine-4-propionic acid

97%

Synonym(s):

3-(2H-1,4-Benzoxazin-3-(4H)-one-4-yl)-propionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H11NO4
CAS Number:
Molecular Weight:
221.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

163-166 °C (lit.)

functional group

carboxylic acid

SMILES string

OC(=O)CCN1C(=O)COc2ccccc12

InChI

1S/C11H11NO4/c13-10-7-16-9-4-2-1-3-8(9)12(10)6-5-11(14)15/h1-4H,5-7H2,(H,14,15)

InChI key

BHULHAHXIGPYKV-UHFFFAOYSA-N

General description

2,5-Dichlorobenzyl bromide, an aryl halide, is an alkylating agent used in chemical synthesis.[1]

Application

2,5-Dichlorobenzyl bromide may be used in the synthesis of methyl 5-(2,5-dichlorobenzyloxy)-2-hydroxybenzoate and methyl 4-(2,5-dichlorobenzyloxy)-2-hydroxybenzoate via chemoselective alkylation of the hydroxyl group of the corresponding ester.[1]

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and bioevaluation of aryl hydroxamates distinguishing between NAD+ and ATP-dependent DNA ligases.
Kukshal V, et al.
MedChemComm, 3(4), 453-461 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service