All Photos(1)
About This Item
Linear Formula:
CF3C6H4CONH2
CAS Number:
Molecular Weight:
189.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
mp
124-127 °C (lit.)
SMILES string
NC(=O)c1cccc(c1)C(F)(F)F
InChI
1S/C8H6F3NO/c9-8(10,11)6-3-1-2-5(4-6)7(12)13/h1-4H,(H2,12,13)
InChI key
XBGXGCOLWCMVOI-UHFFFAOYSA-N
General description
3-(Trifluoromethyl)benzamide can be prepared from 1-bromo-3-trifluoromethylbenzene via palladium-catalyzed aminocarbonylation with formamide. It forms the core moiety of N-[(substituted 1H-imidazol-1-yl)propyl]benzamides, with potential as antihypertensive agents.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Thromboxane synthetase inhibitors and antihypertensive agents. 1. N-[(1H-imidazol-1-yl) alkyl] aryl amides and N-[(1H-1, 2, 4-triazol-1-yl) alkyl] aryl amides.
Wright Jr WB, et al.
Journal of Medicinal Chemistry, 29(4), 523-530 (1986)
Weinreb SM.
Science of Synthesis: Houben-Weyl Methods of Molecular Transformations, 21, 126-126 (2014)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service