Skip to Content
Merck
All Photos(1)

Key Documents

547727

Sigma-Aldrich

1-[(2-Ethylhexyl)oxy]-4-methoxybenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4OCH2CH(C2H5)(CH2)3CH3
CAS Number:
Molecular Weight:
236.35
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

refractive index

n20/D 1.4960 (lit.)

bp

303-304 °C (lit.)

density

0.960 g/mL at 25 °C (lit.)

SMILES string

CCCCC(CC)COc1ccc(OC)cc1

InChI

1S/C15H24O2/c1-4-6-7-13(5-2)12-17-15-10-8-14(16-3)9-11-15/h8-11,13H,4-7,12H2,1-3H3

InChI key

IMRTUSZHEQXAFU-UHFFFAOYSA-N

General description

1-[(2-Ethylhexyl)oxy]-4-methoxybenzene is an intermediate for generating conjugated polymers, which are used in polymer solar cells (PSCs)[1] and light emitting diodes (LED).[2]
Intermediate for a conductive polymer.

Application

1-[(2-Ethylhexyl)oxy]-4-methoxybenzene [4-(2-Ethylheptyloxy)benzaldehyde] may be used in the preparation of 1,4-bis(bromomethyl)-2-((2′-ethylhexyl)oxy)-5-methoxybenzene[1] and 1,4-dibromo-2-[(2-ethylhexyl)oxy]-5-methoxy-benzene.[2]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alternating copolymers of electron-rich arylamine and electron-deficient 2, 1, 3-benzothiadiazole: Synthesis, characterization and photovoltaic properties.
Huo L, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 45(17), 3861-3871 (2007)
Bipolar poly (p-phenylene vinylene)s bearing electron-donating triphenylamine or carbazole and electron-accepting quinoxaline moieties.
Karastatiris P, et al.
Journal of Polymer Science Part A: Polymer Chemistry, 46(7), 2367-2378 (2008)
Poly (quinoxaline vinylene) with conjugated phenylenevinylene side chain: a potential polymer acceptor with broad absorption band.
Huo L, et al.
Macromolecular Chemistry and Physics, 208(12), 1294-1300 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service