Skip to Content
Merck
All Photos(1)

Key Documents

545864

Sigma-Aldrich

(S)-(−)-Aminoglutethimide

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H16N2O2
CAS Number:
Molecular Weight:
232.28
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

optical activity

[α]20/D −160°, c = 1 in methanol

mp

115.5-119.5 °C (lit.)

SMILES string

CC[C@]1(CCC(=O)NC1=O)c2ccc(N)cc2

InChI

1S/C13H16N2O2/c1-2-13(8-7-11(16)15-12(13)17)9-3-5-10(14)6-4-9/h3-6H,2,7-8,14H2,1H3,(H,15,16,17)/t13-/m0/s1

InChI key

ROBVIMPUHSLWNV-ZDUSSCGKSA-N

Gene Information

Application

(S)-(−)-Aminoglutethimide can be used:
  • As an internal standard in the separation of the pyridoglutethimide enantiomers using the HPLC technique.
  • In the study of steroid hormones role on human cytomegalovirus replication in adrenocortical cells.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stereoselective determination of pyridoglutethimide enantiomers in serum with a chiral cellulose-based high-performance liquid chromatographic column using solid phase extraction and UV detection
Hefnawy MM
Journal of Pharmaceutical and Biomedical Analysis, 31(5), 999-1005 (2003)
Marta Trevisan et al.
Journal of cellular physiology, 221(3), 629-641 (2009-08-19)
Following our recent findings on the presence of human cytomegalovirus (HCMV) in the normal human adrenal cortex and in adrenocortical tumors, especially in cortisol-secreting tumors, aim of the present study was to investigate the direct effects of HCMV infection on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service