Skip to Content
Merck
All Photos(1)

Key Documents

PHR3173

Supelco

Aminoglutethimide

pharmaceutical secondary standard, certified reference material

Synonym(s):

DL-Aminoglutethimide, 3-(4-Aminophenyl)-3-ethyl-2,6-piperidinedione, 3-(p-Aminophenyl)-3-ethylpiperidine-2,6-dione

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$345.00

$345.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
500 MG
$345.00

About This Item

Empirical Formula (Hill Notation):
C13H16N2O2
CAS Number:
Molecular Weight:
232.28
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

$345.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

grade

certified reference material
pharmaceutical secondary standard

Quality Level

Agency

traceable to BP BP684
traceable to USP 1025205

API family

aminoglutethimide

CofA

current certificate can be downloaded

packaging

pkg of 500 mg

mp

152-154 °C (lit.)

application(s)

pharmaceutical

storage temp.

2-30°C

SMILES string

CCC1(CCC(=O)NC1=O)c2ccc(N)cc2

InChI

1S/C13H16N2O2/c1-2-13(8-7-11(16)15-12(13)17)9-3-5-10(14)6-4-9/h3-6H,2,7-8,14H2,1H3,(H,15,16,17)

InChI key

ROBVIMPUHSLWNV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Application

These Secondary Standards are qualified as Certified Reference Materials. These are suitable for use in several analytical applications including but not limited to pharma release testing, pharma method development for qualitative and quantitative analyses, food and beverage quality control testing, and other calibration requirements.

Biochem/physiol Actions

DL-Aminoglutethimide is a derivative of the sedative glutethimide. Originally introduced as an anticonvulsant, it was found to cause adrenal insufficiency. Blocks adrenal steroidogenesis by inhibiting the enzymatic conversion of cholesterol to pregnenolone. It also blocks the peripheral conversion (aromatization) of androgenic precursors to estrogens. The D-isomer is 30 times more potent at inhibiting aromatase activity, whereas the L-isomer is more potent at inhibiting cholesterol side-chain cleavage (steroidogenesis).
Derivative of the sedative glutethimide. Originally introduced as an anticonvulsant, it was found to cause adrenal insufficiency. Blocks adrenal steroidogenesis by inhibiting the enzymatic conversion of cholesterol to pregnenolone.

Analysis Note

These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.

Other Notes

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Footnote

To see an example of a Certificate of Analysis for this material enter LRAC8223 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service