Skip to Content
Merck
All Photos(1)

Key Documents

544612

Sigma-Aldrich

3-Ethyl-2-pentene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH=C(CH2CH3)2
CAS Number:
Molecular Weight:
98.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.414 (lit.)

bp

94 °C (lit.)

density

0.717 g/mL at 25 °C (lit.)

SMILES string

CC\C(CC)=C/C

InChI

1S/C7H14/c1-4-7(5-2)6-3/h4H,5-6H2,1-3H3

InChI key

XMYFZAWUNVHVGI-UHFFFAOYSA-N

General description

3-Ethyl-2-pentene is a trisubstituted olefin.

Application

3-Ethyl-2-pentene may be used in the preparation of:
  • (2E,3R)-2-hydroxy-3,4-dimethyl-N-phenylpent-4-enamide[1]
  • 3-ethyl-1-pentene[2]
  • 5-ethyl-2,4-dimethyl-4-hepten-3-one[3]

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

36.0 °F - closed cup

Flash Point(C)

2.2 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Spectral correlations for ?,?-unsaturated ketones.
Faulk DD and Fry A.
The Journal of Organic Chemistry, 35(2), 364-369 (1970)
Enantioselective syn-selective scandium-catalyzed ene reactions.
Evans DA and Wu J.
Journal of the American Chemical Society, 127(22), 8006-8007 (2005)
Organoboranes. IV. The displacement reaction with organoboranes derived from the hydroboration of branched-chain olefins. A contrathermodynamic isomerization of olefins.
Brown HC and Bhatt MV.
Journal of the American Chemical Society, 88(7), 1440-1443 (1966)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service