Skip to Content
Merck
All Photos(1)

Key Documents

542814

Sigma-Aldrich

3-Pyridylamidoxime

98%

Synonym(s):

N-Hydroxynicotinamidine

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$65.80

$65.80


Estimated to ship onApril 16, 2025


Request a Bulk Order

Select a Size

Change View
1 G
$65.80

About This Item

Empirical Formula (Hill Notation):
C6H7N3O
CAS Number:
Molecular Weight:
137.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$65.80


Estimated to ship onApril 16, 2025


Request a Bulk Order

Quality Level

Assay

98%

mp

130-135 °C (lit.)

functional group

amine
oxime

SMILES string

N\C(=N\O)c1cccnc1

InChI

1S/C6H7N3O/c7-6(9-10)5-2-1-3-8-4-5/h1-4,10H,(H2,7,9)

InChI key

AQBMQGDKWIPBRF-UHFFFAOYSA-N

General description

3-Pyridylamidoxime (pyridine-3-amidoxime, Py-3-AO) can be synthesized from the reaction between hydroxylamine hydrochloride, sodium hydroxide and 3-cyanopyridine (dissolved in ethanol).[1]

Application

3-Pyridylamidoxime may be used in the synthesis of 3-(3-pyridyl)-5-trichloromethyl-1,2,4-oxadiazole.[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Separation of isomeric monocyanopyridine and pyridinemonoamidoxime hydrochlorides by thin-layer chromatography.
Pearse Jr GA.
Journal of Chromatography A, 177, 174-176 (1979)
Studies on the Synthese of N-Heterocyclic Compounds. III. Hypocholesterolemic 1, 2, 4-Oxadiazole Derivatives (1).
Yurugi S, et al.
Chemical & Pharmaceutical Bulletin, 21(8), 1641-1650 (1973)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service