Skip to Content
Merck
All Photos(1)

Key Documents

242632

Sigma-Aldrich

2-Chloroethyl methyl sulfide

97%

Synonym(s):

2-(Methylthio)ethyl chloride, Hemisulfur mustard

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$146.00

$146.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$146.00

About This Item

Linear Formula:
ClCH2CH2SCH3
CAS Number:
Molecular Weight:
110.61
Beilstein:
1731033
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$146.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.496 (lit.)

bp

55-56 °C/30 mmHg (lit.)

density

1.11 g/mL at 25 °C (lit.)

functional group

chloro
thioether

storage temp.

2-8°C

SMILES string

CSCCCl

InChI

1S/C3H7ClS/c1-5-3-2-4/h2-3H2,1H3

InChI key

MYFKLQFBFSHBPA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

2-Chloroethyl methyl sulfide is a monofunctional chemical analog of sulfur mustard and has been used to study the potential of 2,6-dithiopurine as an antagonist of sulfur mustard[1].

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1A - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

107.6 °F - closed cup

Flash Point(C)

42 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

G R Klinefelter et al.
Journal of andrology, 18(2), 139-150 (1997-03-01)
In a previous study, we found that ethane dimethanesulphonate (EDS) compromised the fertilizing ability of proximal cauda epididymal sperm from the rat within 4 days of exposure, an effect that persisted in castrated, testosterone (T)-implanted animals, establishing direct action on
Y W Sohn et al.
Carcinogenesis, 17(6), 1361-1364 (1996-06-01)
Vinthionine (S-vinyl-DL-homocysteine) is hepatocarcinogenic in rats and mice. [Vinyl-14C]vinthionine binds covalently to rat liver DNA, RNA and protein in vivo, but not in vitro. This amino acid is directly mutagenic in Salmonella typhimurium TA100 and TA1535; the mechanism of its
J R King et al.
Toxicology and applied pharmacology, 104(1), 167-179 (1990-06-01)
Previous research has shown the isolated perfused porcine skin flap (IPPSF) to be a novel in vitro experimental model for investigating xenobiotic percutaneous absorption. In this study, the IPPSF was used to biochemically and morphologically assess the dermatotoxicity of 2-chloroethyl
K Leslie Powell et al.
Chemical research in toxicology, 23(3), 497-503 (2010-01-07)
Sulfur mustard (bis-(2-chloroethyl)sulfide) is a well-known chemical warfare agent that induces debilitating cutaneous toxicity in exposed individuals. It is also known to be carcinogenic and mutagenic because of its ability to damage DNA via electrophilic attack. We previously showed that
Bruna Scaggiante et al.
Hematological oncology, 31(2), 110-116 (2012-08-30)
The elongation factor 1A proteins (eEF1A1/A2) are known to play a role in tumours. We previously found that a more basic isoform of eEF1A (MBI-eEF1A) is present in the cytoskeletal/nuclear-enriched extracts of CCRF-CEM T-lymphoblasts but not in those of normal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service