Skip to Content
Merck
All Photos(1)

Key Documents

541753

Sigma-Aldrich

3,4-Dichloroiodobenzene

98%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$130.00

$130.00


Estimated to ship onApril 12, 2025


Request a Bulk Order

Select a Size

Change View
25 G
$130.00

About This Item

Linear Formula:
Cl2C6H3I
CAS Number:
Molecular Weight:
272.90
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$130.00


Estimated to ship onApril 12, 2025


Request a Bulk Order

Quality Level

Assay

98%

mp

27-29 °C (lit.)

functional group

chloro
iodo

SMILES string

Clc1ccc(I)cc1Cl

InChI

1S/C6H3Cl2I/c7-5-2-1-4(9)3-6(5)8/h1-3H

InChI key

NADPFZNWCQIJJW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3,4-Dichloroiodobenzene undergoes Stille coupling reaction with vinyltributyltin in the presence of Pd(0)precursors to furnish styrene derivative.[1]

Application

3,4-Dichloroiodobenzene may be used in the preparation of:
  • tetrachloromethoxybiphenyls[2]
  • 1-(3,4-dichlorophenyl)-2-trimethylsilylacetylene[3]
  • 2-(3,4-dichlorocinnamyl)isoindoline-1,3-dione[4]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of chlorinated methoxybiphenyls as contaminants in fish and as potential interferences in the determination of chlorinated dibenzo-p-dioxins.
D W Phillipson et al.
Analytical chemistry, 52(14), 2328-2332 (1980-12-01)
Synthesis and properties of various poly (diphenylacetylenes) containing tert-amine moieties.
Katsumata T, et al.
Polymer, 49(12), 2808-2816 (2008)
Leonardo S Santos et al.
The Journal of organic chemistry, 72(15), 5809-5812 (2007-06-26)
On-line monitoring of Stille reactions was performed via direct infusion electrospray ionization mass spectrometry (ESI-MS) and its tandem version (ESI-MS/MS). When operated in the positive ion mode, ESI(+)-MS was able to transfer, directly from solution to the gas phase, the
Cara A Mosley et al.
Bioorganic & medicinal chemistry, 17(17), 6463-6480 (2009-08-04)
The synthesis and structure-activity relationship analysis of a novel class of amide-based biaryl NR2B-selective NMDA receptor antagonists are presented. Some of the studied compounds are potent, selective, non-competitive, and voltage-independent antagonists of NR2B-containing NMDA receptors. Like the founding member of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service