Skip to Content
Merck
All Photos(1)

Key Documents

539449

Sigma-Aldrich

1-(N-Boc-aminomethyl)-4-(aminomethyl)benzene

95%

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$105.00
5 G
$445.00

$105.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
1 G
$105.00
5 G
$445.00

About This Item

Linear Formula:
H2NCH2C6H4CH2NHCO2C(CH3)3
CAS Number:
Molecular Weight:
236.31
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$105.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

95%

mp

64-68 °C (lit.)

functional group

amine

SMILES string

CC(C)(C)OC(=O)NCc1ccc(CN)cc1

InChI

1S/C13H20N2O2/c1-13(2,3)17-12(16)15-9-11-6-4-10(8-14)5-7-11/h4-7H,8-9,14H2,1-3H3,(H,15,16)

InChI key

NUANLVJLUYWSER-UHFFFAOYSA-N

Related Categories

General description

1-(N-Boc-aminomethyl)-4-(aminomethyl)benzene is a protected amine.

Application

1-(N-Boc-aminomethyl)-4-(aminomethyl)benzene may be used for the synthesis of:
  • oligomeric thioureas[1]
  • bipyrrole-based[2]catenane[2]
  • model receptors for dicarboxylates and monosaccharides[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Andrievsky A, et al.
Journal of the American Chemical Society, 120, 9712-9712 (1998)
Benito JM, et al.
The Journal of Organic Chemistry, 66, 1366-1366 (2001)
Smith J, et al.
The Journal of Organic Chemistry, 61, 8811-8811 (1996)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service