Skip to Content
Merck
All Photos(1)

Key Documents

538523

Sigma-Aldrich

6-Bromo-1,4-benzodioxane

97%

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$62.50

$62.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$62.50

About This Item

Empirical Formula (Hill Notation):
C8H7BrO2
CAS Number:
Molecular Weight:
215.04
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$62.50

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

refractive index

n20/D 1.588 (lit.)

bp

259-260 °C (lit.)

density

1.598 g/mL at 25 °C (lit.)

functional group

bromo

SMILES string

Brc1ccc2OCCOc2c1

InChI

1S/C8H7BrO2/c9-6-1-2-7-8(5-6)11-4-3-10-7/h1-2,5H,3-4H2

InChI key

LFCURAJBHDNUNG-UHFFFAOYSA-N

General description

6-Bromo-1,4-benzodioxane is an aryl halide. It can be synthesized from 1,4-benzodioxane, via bromination with bromine in acetic acid. It undergoes alkoxycarbonylation reaction with sodium tert-butoxide in the presence of CO to yield 2,3-dihydro-benzo[1,4]dioxine-6-carboxylic acid t-butyl ester.[1]

Application

6-Bromo-1,4-benzodioxane may be used as a starting reagent in the synthesis of chiral diphosphines (SYNPHOS and DIFLUORPHOS).[2]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chiral biphenyl diphosphines for asymmetric catalysis: Stereoelectronic design and industrial perspectives.
Jeulin, Severine, et al.
Proceedings of the National Academy of Sciences of the USA, 101(16), 5799-5804 (2004)
An Efficient Method for the Preparation of Tertiary Esters by Palladium-Catalyzed Alkoxycarbonylation of Aryl Bromides.
Xin, Zhuo, et al.
Organic Letters, 14(1), 284-287 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service