Select a Size
$448.92
List Price$522.00Save 14%Select a Size
About This Item
$448.92
List Price$522.00Save 14%Recommended Products
Assay
97%
refractive index
n20/D 1.508 (lit.)
bp
84 °C/6 mmHg (lit.)
density
0.958 g/mL at 25 °C (lit.)
SMILES string
CCc1ccc(OC)cc1
1 of 4
This Item | SAB5200988 | SAB5200888 | SAB5200904 |
---|---|---|---|
conjugate unconjugated | conjugate Allophycocyanin conjugate | conjugate unconjugated | conjugate phycoerythrin (R-PE) conjugate |
species reactivity mouse, human, rat | species reactivity mouse, human, rat | species reactivity mouse, human, rat | species reactivity human, mouse, rat |
Gene Information rat ... Grin2a(24409) | Gene Information rat ... Grin2a(24409) | Gene Information rat ... Grin2a(24409) | Gene Information rat ... Grin2a(24409) |
clone S327A-38, monoclonal | clone S327A-38, monoclonal | clone S327-95, monoclonal | clone S327-95, monoclonal |
antibody form purified immunoglobulin | antibody form purified immunoglobulin | antibody form purified immunoglobulin | antibody form purified immunoglobulin |
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
174.9 °F - closed cup
Flash Point(C)
79.4 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service