Skip to Content
Merck
All Photos(1)

Key Documents

532215

Sigma-Aldrich

N-Methyl-3-chloroaniline

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4NHCH3
CAS Number:
Molecular Weight:
141.60
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.5840 (lit.)

bp

243-244 °C (lit.)

density

1.156 g/mL at 25 °C (lit.)

functional group

amine
chloro

SMILES string

CNc1cccc(Cl)c1

InChI

1S/C7H8ClN/c1-9-7-4-2-3-6(8)5-7/h2-5,9H,1H3

InChI key

WFGYSQDPURFIFL-UHFFFAOYSA-N

General description

N-Methyl-3-chloroaniline, also known as 3-chloro-N-methylaniline, is a secondary amine. It can undergo C3 borylation in the presence of an iridium catalyst.[1]

Application

N-Methyl-3-chloroaniline may be used as a starting material in the preparation of:2
  • N

  • -(2-bromoethyl)-N-methyl-3-chloroaniline
  • 2-[2-(N-methyl-3-chloroanilino)ethyl]-1,3-isoindolinedione
  • N1-(3-chlorophenyl)-N1-methyl-1,2-ethanediamine dihydrochloride

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

>230.0 °F - closed cup

Flash Point(C)

> 110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Effect of modifications of the alkylpiperazine moiety of trazodone on 5HT2A and alpha1 receptor binding affinity.
Giannangeli, M
Medicinal Chemistry, 42 (3), 336-345 (1999)
A Traceless Directing Group for C-H Borylation.
Preshlock SM
Angewandte Chemie (Weinheim an der Bergstrasse, Germany), 125(49), 13153-13157 (2013)
M Giannangeli et al.
Journal of medicinal chemistry, 42(3), 336-345 (1999-02-13)
A series of triazolopyridine derivatives (compounds 2a-l) were synthesized in order to explore the effect of modifications of the alkylpiperazine moiety of trazodone (fragment A) on binding affinity for 5HT2A and alpha1 receptors. All of the synthesized compounds show a

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service