All Photos(1)
About This Item
Linear Formula:
BrC6H3(F)NH2
CAS Number:
Molecular Weight:
190.01
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
97%
mp
43-47 °C (lit.)
SMILES string
Nc1cc(F)ccc1Br
InChI
1S/C6H5BrFN/c7-5-2-1-4(8)3-6(5)9/h1-3H,9H2
InChI key
FWTXFEKVHSFTDQ-UHFFFAOYSA-N
Application
2-Bromo-5-fluoroaniline may be used to synthesize:
- orthobromothiobenzanilide of 5-fluoronitrobenzothiazole
- 2-bromo-5-fluorobenzo[14C]nitrile
- [3-14C]-5-fluoro-1,3-dihydro-1-hydroxy-2,1-benzoxaborole
- 2-bromo-5-fluoro-phenyl)-(2-methoxy-6-pentadecyl-benzyl)amine
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
230.0 °F - closed cup
Flash Point(C)
110 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Synthesis of novel benzylamine analogues of Anacardic acid as potent antibacterial agents.
Dubey PK.
Der Pharma Chemica, 3(6), 500-512 (2011)
T D Bradshaw et al.
Current medicinal chemistry, 11(8), 1009-1021 (2004-04-14)
This review traces the development of a series of potent and selective antitumour benzothiazoles from the discovery of the initial lead compound, 2-(4-amino-3-methylphenyl)benzothiazole (DF 203) in 1995 to the identification of a clinical candidate, Phortress, scheduled to enter Phase 1
Synthesis of isotopically labelled [3-14C]-and [3, 3-2H2]-5-fluoro-1, 3-dihydro-1-hydroxy-2, 1-benzoxaborole (AN2690), a new antifungal agent for the potential treatment of onychomycosis.
Baker SJ, et al.
Journal of Labelled Compounds & Radiopharmaceuticals, 50(4), 245-250 (2007)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service