Skip to Content
Merck
All Photos(3)

Key Documents

F3800

Sigma-Aldrich

4-Fluoroaniline

99%

Synonym(s):

1-Amino-4-fluorobenzene

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$57.60
100 G
$112.00
500 G
$470.00

$57.60

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 21, 2025Details


Request a Bulk Order

Select a Size

Change View
25 G
$57.60
100 G
$112.00
500 G
$470.00

About This Item

Linear Formula:
FC6H4NH2
CAS Number:
Molecular Weight:
111.12
Beilstein:
742030
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$57.60

PRICE WITHOUT NATIONAL TAXES

Available to ship onApril 21, 2025Details


Request a Bulk Order

Assay

99%

form

liquid

refractive index

n20/D 1.539 (lit.)

bp

187 °C/767 mmHg (lit.)

density

1.173 g/mL at 25 °C (lit.)

SMILES string

Nc1ccc(F)cc1

InChI

1S/C6H6FN/c7-5-1-3-6(8)4-2-5/h1-4H,8H2

InChI key

KRZCOLNOCZKSDF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1C - STOT RE 2

Target Organs

Blood,hematopoietic system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

162.9 °F

Flash Point(C)

72.7 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C V Eadsforth et al.
Xenobiotica; the fate of foreign compounds in biological systems, 16(6), 555-566 (1986-06-01)
o-Fluoroaniline is rapidly metabolized and excreted in rats, rabbits and marmosets. Following a single oral dose of 14C-fluoroaniline of about 20 mg/kg, more than 80% of the dose is excreted in 0-24 h, the urine being the major route of
Meizhen Wang et al.
Environmental science and pollution research international, 20(9), 6201-6209 (2013-04-17)
To develop a bacterial bioaugmentation system for fluorine-containing industrial wastewater treatment, optimal conditions for 4-fluoroaniline (4-FA) degradation and autoinducer release in Acinetobacter sp. TW were determined. Quorum sensing in biofilms of strain TW was also investigated. Different optimal conditions exist
G B Scarfe et al.
Xenobiotica; the fate of foreign compounds in biological systems, 29(2), 205-216 (1999-04-13)
1. The urinary metabolic fate of 4-fluoroaniline (4-FA) and 1-[13C]-4-fluoroacetanilide (4-FAA) has been studied using NMR-based methods after 50 and 100 mg kg(-1) i.p. doses respectively to the male Sprague-Dawley rat. 2. 4-FA was both ortho- and para-hydroxylated. The major
I M Rietjens et al.
Chemico-biological interactions, 77(3), 263-281 (1991-01-01)
Metabolism and bioactivation of fluoroanilines was studied both in vitro in microsomal systems and in vivo. 4-Fluoroaniline and pentafluoroaniline and their non-para fluorinated analogues were used as the model compounds. Special attention was focussed on bioactivation to reactive benzoquinoneimines. Cytochrome
J G Bundy et al.
Xenobiotica; the fate of foreign compounds in biological systems, 32(6), 479-490 (2002-08-06)
1. Little is known about metabolism of xenobiotics by earthworms, despite their importance in soil ecotoxicity testing. Normal earthworms and earthworms treated with antibiotics to ensure inhibition of gut microflora were exposed to two model xenobiotic compounds, 4-fluoroaniline and 4-fluorobiphenyl

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service