Skip to Content
Merck
All Photos(2)

Key Documents

528331

Sigma-Aldrich

N-Methyl-N-(2-hydroxyethyl)-4-aminobenzaldehyde

96%

Sign Into View Organizational & Contract Pricing

Select a Size

25 G
$190.00

$190.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
25 G
$190.00

About This Item

Linear Formula:
(HOCH2CH2)(CH3)NC6H4CHO
CAS Number:
Molecular Weight:
179.22
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$190.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Quality Level

Assay

96%

mp

66-70 °C (lit.)

functional group

aldehyde
amine
hydroxyl

SMILES string

CN(CCO)c1ccc(C=O)cc1

InChI

1S/C10H13NO2/c1-11(6-7-12)10-4-2-9(8-13)3-5-10/h2-5,8,12H,6-7H2,1H3

InChI key

JOCUIVLSLBBESN-UHFFFAOYSA-N

Application

N-Methyl-N-(2-hydroxyethyl)-4-aminobenzaldehyde may be used to synthesize:
  • 2,5-bis[4-(methylaminoethanol)benzylidene]cyclopentanone[1]
  • 4′-[(2-tosyloxyethyl)(methyl)amino]-4-phenyl-3-buten-2-malonitrile, a tosylate precursor[2]
  • methanesulfonic acid 2-[(4-formyl-phenyl)-methyl-amino]-ethyl ester[3]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Synthesis and characterization of dendritic nonlinear optical chromophore containing phenylene attached with bulky alkyl group.
Choi JJ, et al.
Macromolecular Research, 15(1), 59-64 (2007)
18F-FBM: A new PET radiotracer for imaging ?-amyloid plaques and neurofibrillary tangles.
Wang C, et al.
Journal of Nuclear Medicine, 47(1), 217P-217P (2006)
Waqas Ahmad et al.
Chemical biology & drug design, 83(6), 670-681 (2014-01-11)
Arachidonic acid and its metabolites have generated a heightened interest due to their significant role in inflammation. Inhibiting the enzymes involved in arachidonic acid metabolism has been considered as the synergistic anti-inflammatory effect. A series of novel curcumin diarylpentanoid analogues

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service