Recommended Products
Assay
96%
mp
68-71 °C (lit.)
SMILES string
OCc1cc(ccc1F)[N+]([O-])=O
InChI
1S/C7H6FNO3/c8-7-2-1-6(9(11)12)3-5(7)4-10/h1-3,10H,4H2
InChI key
IFIOUOYJVOSTFH-UHFFFAOYSA-N
General description
2-Fluoro-5-nitrobenzyl alcohol can be prepared from 2-fluoro-5-nitrobenzaldehyde.
Application
2-Fluoro-5-nitrobenzyl alcohol may be used to synthesize 2-fluoro-5-nitrobenzyl bromide and 5-amino-2-fluorobenzyl alcohol.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Chemical & pharmaceutical bulletin, 64(6), 594-601 (2016-06-03)
Malaria is one of the most important tropical diseases; the use of amodiaquine as a current chemotherapy in the treatment of malaria has shown some problems such as hepatotoxicity and agranulocytosis. In this work we present the rational design, synthesis
4H-1-Benzopyrans by a tandem SN2-SNAr reaction.
Journal of Heterocyclic Chemistry, 45(2), 547- 550 (2008)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service