525855
Diethyl allyl phosphate
98%
Synonym(s):
Allyl ethyl phosphate, Diethyl 2-propen-1-yl phosphate
Select a Size
Select a Size
About This Item
Recommended Products
Assay
98%
refractive index
n20/D 1.422 (lit.)
bp
45-46 °C (lit.)
density
1.09 g/mL at 25 °C (lit.)
functional group
phosphate
SMILES string
CCOP(=O)(OCC)OCC=C
InChI
1S/C7H15O4P/c1-4-7-11-12(8,9-5-2)10-6-3/h4H,1,5-7H2,2-3H3
InChI key
GZNJJEODYYLYSA-UHFFFAOYSA-N
General description
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
230.0 °F - closed cup
Flash Point(C)
110 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service