Skip to Content
Merck
All Photos(1)

Key Documents

522392

Sigma-Aldrich

Cobalt(II) cyanide dihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Co(CN)2 · 2H2O
CAS Number:
Molecular Weight:
147.00
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
grade:
for analytical purposes
form:
powder

grade

for analytical purposes

form

powder

SMILES string

O.O.N#C[Co]C#N

InChI

1S/2CN.Co.2H2O/c2*1-2;;;/h;;;2*1H2

InChI key

BQEYQDMFPKMUSG-UHFFFAOYSA-N

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service