Skip to Content
Merck
All Photos(1)

Key Documents

520470

Sigma-Aldrich

2-(Ethoxycarbonyl)phenylzinc bromide solution

0.5 M in THF

Sign Into View Organizational & Contract Pricing

Select a Size

50 ML
$408.00

$408.00


Estimated to ship onMay 24, 2025


Request a Bulk Order

Select a Size

Change View
50 ML
$408.00

About This Item

Linear Formula:
C2H5O2CC6H4ZnBr
CAS Number:
Molecular Weight:
294.46
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

$408.00


Estimated to ship onMay 24, 2025


Request a Bulk Order

concentration

0.5 M in THF

density

0.992 g/mL at 25 °C

functional group

ester

storage temp.

2-8°C

SMILES string

CCOC(=O)c1ccccc1[Zn]Br

InChI

1S/C9H9O2.BrH.Zn/c1-2-11-9(10)8-6-4-3-5-7-8;;/h3-6H,2H2,1H3;1H;/q;;+1/p-1

InChI key

XOFOTLKSCXXHTH-UHFFFAOYSA-M

Legal Information

Product of Rieke Metals, Inc.
Rieke is a registered trademark of Rieke Metals, Inc.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service