Skip to Content
Merck
All Photos(1)

Key Documents

518700

Sigma-Aldrich

2-Bromo-4-(trifluoromethyl)aniline

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3(CF3)NH2
CAS Number:
Molecular Weight:
240.02
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.524 (lit.)

bp

109-110 °C/10 mmHg (lit.)

SMILES string

Nc1ccc(cc1Br)C(F)(F)F

InChI

1S/C7H5BrF3N/c8-5-3-4(7(9,10)11)1-2-6(5)12/h1-3H,12H2

InChI key

QKRJIXSZTKOFTD-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G B Scarfe et al.
Xenobiotica; the fate of foreign compounds in biological systems, 28(4), 373-388 (1998-05-30)
1. The metabolic fate and urinary excretion of 2-bromo-4-trifluoromethylaniline has been studied in rat using 19F-NMR spectroscopic and directly coupled HPLC-NMR-MS methods. The compound was dosed to Sprague-Dawley rats (50 mg kg-1, i.p.) and urine collected over 0-8, 8-24 and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service