Skip to Content
Merck
All Photos(2)

Documents

514071

Sigma-Aldrich

Copper(II) tartrate hydrate

99.9% trace metals basis

Synonym(s):

Cupric tartrate hydrate, Tartaric acid cupric salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-CH(OH)CO2]2Cu·xH2O
CAS Number:
Molecular Weight:
211.62 (anhydrous basis)
Beilstein:
4166088
EC Number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

99.9% trace metals basis

form

solid

impurities

≤1500.0 ppm Trace Metal Analysis

mp

~275 °C (dec.) (lit.)

application(s)

battery manufacturing

SMILES string

O.[Cu++].O[C@H]([C@@H](O)C([O-])=O)C([O-])=O

InChI

1S/C4H6O6.Cu.H2O/c5-1(3(7)8)2(6)4(9)10;;/h1-2,5-6H,(H,7,8)(H,9,10);;1H2/q;+2;/p-2/t1-,2-;;/m1../s1

InChI key

MAUZTCHAIPUZJB-OLXYHTOASA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Copper tartrate may be used as a catalyst for the fabrication of helical polyacetylene fibers by the polymerization of acetylene.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Symmetric and helical growth of polyacetylene fibers over a single copper crystal derived from copper tartrate decomposition.
Qin Y, et al.
Organic Letters, 4(18), 3123-3125 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service