Skip to Content
Merck
All Photos(1)

Key Documents

513121

Sigma-Aldrich

sec-Butyl ether

96%

Synonym(s):

Di-sec-butyl ether

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$251.00

$251.00


Estimated to ship onMarch 26, 2025


Request a Bulk Order

Select a Size

Change View
5 G
$251.00

About This Item

Linear Formula:
CH3CH2CH(CH3)OCH(CH3)CH2CH3
CAS Number:
Molecular Weight:
130.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay:
96%

$251.00


Estimated to ship onMarch 26, 2025


Request a Bulk Order

Quality Level

Assay

96%

refractive index

n20/D 1.394 (lit.)

bp

121 °C (lit.)

density

0.759 g/mL at 25 °C (lit.)

functional group

ether

SMILES string

CCC(C)OC(C)CC

InChI

1S/C8H18O/c1-5-7(3)9-8(4)6-2/h7-8H,5-6H2,1-4H3

InChI key

HHBZZTKMMLDNDN-UHFFFAOYSA-N

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

78.1 °F - closed cup

Flash Point(C)

25.6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vapor-liquid and liquid-liquid equilibria for the system sec-butyl-alcohol-di-sec-butyl ether-water.
Kovach III JW and Seider WD.
Journal of Chemical and Engineering Data, 33(1), 16-20 (1988)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service