Recommended Products
Assay
95%
refractive index
n20/D 1.427 (lit.)
bp
165 °C (lit.)
density
0.837 g/mL at 25 °C (lit.)
SMILES string
C=CCCCC#N
InChI
1S/C6H9N/c1-2-3-4-5-6-7/h2H,1,3-5H2
InChI key
UNAQSRLBVVDYGP-UHFFFAOYSA-N
General description
5-Hexenenitrile, an aliphatic nitrile, is a volatile compound that is formed as a degradation product of glucobrassicanapin in Aurinia sinuata. It is reported to be an attractant from oilseed rape for the cabbage seed weevil, Ceutorhynchus assimilis. 5-Hexenenitrile is the main volatile compound found in wheat extract. It undergoes Wacker oxidation to form the corresponding methyl ketone using PdCl2-DMA (palladium dichloride-N,N-diemthylacetamide) catalytic system.
Application
5-Hexenenitrile may be used in the synthesis of 1,8-dicyano-4-octene.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
104.9 °F - closed cup
Flash Point(C)
40.5 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Composition of the young green barley and wheat leaves.
J. Essent. Oil Res., 19(2), 134-137 (2007)
Convenient and Efficient Pd-Catalyzed Regioselective Oxyfunctionalization of Terminal Olefins by Using Molecular Oxygen as Sole Reoxidant.
Angewandte Chemie (Weinheim an der Bergstrasse, Germany), 118(3), 495-499 (2006)
Polymer, 49(24), 5307-5313 (2009-11-13)
High molecular weight, high functionality diamino telechelic polybutadienes (TPBs) were synthesized by ring-opening metathesis polymerization (ROMP) of 1,5-cyclooctadiene (COD) in the presence of a chain transfer agent, 1,8-dicyano-4-octene, followed by lithium aluminum hydride reduction. Melt coupling of diamino TPB with
The responses of the cabbage seed weevil Ceutorhynchus assimilis to volatile compounds from oilseed rape in a linear track olfactometer.
Entomologia Experimentalis et Applicata, 85(3), 257-262 (1997)
Glucosinolates, glycosidically bound volatiles and antimicrobial activity of Aurinia sinuata (Brassicaceae).
Food Chemistry, 121(4), 1020-1028 (2010)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service