Skip to Content
Merck
All Photos(1)

Key Documents

51092

Sigma-Aldrich

3,4,5-Trifluorophenol

≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H3F3O
CAS Number:
Molecular Weight:
148.08
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0%

form

solid

mp

52-55 °C

functional group

fluoro

SMILES string

Oc1cc(F)c(F)c(F)c1

InChI

1S/C6H3F3O/c7-4-1-3(10)2-5(8)6(4)9/h1-2,10H

InChI key

ZRTWIJKGTUGZJY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3,4,5-Trifluorophenol is a halo-substituted phenol. It participates in the synthesis of difluorooxymethylene-bridged liquid crystals.[1]

Application

3,4,5-Trifluorophenol may be used in the synthesis of 3,4,5-trifluorophenoxymethyl-substituted polystyrene.[2]

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Difluorooxymethylene-Bridged Liquid Crystals: A Novel Synthesis Based on the Oxidative Alkoxydifluorodesulfuration of Dithianylium Salts.
Kirsch P, et al.
Angewandte Chemie (International Edition in English), 40(8), 1480-1484 (2001)
Liquid crystal alignment properties of polystyrene derivatives containing fluorinated side groups.
Kang H, et al.
Macromolecular Research, 18(1), 78-85 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service