Skip to Content
Merck
All Photos(1)

Key Documents

196789

Sigma-Aldrich

2,3,5,6-Tetrafluorophenol

97%

Synonym(s):

1,2,4,5-Tetrafluoro-3-hydroxybenzene

Sign Into View Organizational & Contract Pricing

Select a Size

5 G
$68.90
25 G
$228.00

$68.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
5 G
$68.90
25 G
$228.00

About This Item

Linear Formula:
HC6F4OH
CAS Number:
Molecular Weight:
166.07
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$68.90

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

97%

form

solid

bp

140 °C (lit.)

mp

37-39 °C (lit.)

functional group

fluoro

SMILES string

Oc1c(F)c(F)cc(F)c1F

InChI

1S/C6H2F4O/c7-2-1-3(8)5(10)6(11)4(2)9/h1,11H

InChI key

PBYIIRLNRCVTMQ-UHFFFAOYSA-N

Application

2,3,5,6-Tetrafluorophenol was used in preparation of :
  • radioiodinated phenylalanine derivative which is useful in peptide synthesis[1]
  • technetium-99m labeled antibodies[2]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

A R Fritzberg et al.
Proceedings of the National Academy of Sciences of the United States of America, 85(11), 4025-4029 (1988-06-01)
Technetium-99m labeling of antibodies has been suboptimal because of low affinity adventitious binding, nonspecific labeling, and loss of immunoreactivity. The diamide dithiolate ligand system (N2S2) forms highly stable, well-defined tetradentate complexes with Tc(V). Antibodies and their fragments have been labeled
K Detmer et al.
The Journal of biological chemistry, 260(10), 5998-6005 (1985-05-25)
The oxidative half-reaction of phenol hydroxylase has been studied by stopped-flow spectrophotometry. Three flavin-oxygen intermediates can be detected when the substrate is thiophenol, or m-NH2, m-OH, m-CH3, m-Cl, or p-OH phenol. Intermediate I, the flavin C(4a)-hydroperoxide, has an absorbance maximum
D S Wilbur et al.
Bioconjugate chemistry, 4(6), 574-580 (1993-11-01)
An investigation to prepare a phenylalanine derivative which could be radioiodinated and used directly in peptide synthesis was conducted. N-Boc-p-(tri-n-butylstannyl)-L-phenylalanine tetrafluorophenyl ester was targeted and synthesized from N-Boc-p-iodo-L-phenylalanine. The requisite aryl stannylation reaction was found to be best conducted using
G A Eiceman et al.
Journal of the American Society for Mass Spectrometry, 10(11), 1157-1165 (2001-09-07)
Atmospheric pressure chemical ionization (APCI)-mass spectrometry (MS) for fluorinated phenols (C6H5-xFxOH Where x = 0-5) in nitrogen with Cl- as the reagent ion yielded product ions of M Cl- through ion associations or (M-H)- through proton abstractions. Proton abstraction was
C den Besten et al.
Chemical research in toxicology, 6(5), 674-680 (1993-09-01)
In the present study the oxidative dehalogenation of a para-halogenated phenol was studied using pentafluorophenol and its non-para-halogenated analogue 2,3,5,6-tetrafluorophenol as model compounds. 19F NMR was used to characterize the metabolite patterns. In order to study the primary oxidation products

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service