Skip to Content
Merck
All Photos(1)

Key Documents

510122

Sigma-Aldrich

3-(Trifluoromethoxy)phenylboronic acid

≥95%

Synonym(s):

3-Trifluoromethoxybenzeneboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3OC6H4B(OH)2
CAS Number:
Molecular Weight:
205.93
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95%

mp

84-89 °C (lit.)

SMILES string

OB(O)c1cccc(OC(F)(F)F)c1

InChI

1S/C7H6BF3O3/c9-7(10,11)14-6-3-1-2-5(4-6)8(12)13/h1-4,12-13H

InChI key

UWDFWVLAHRQSKK-UHFFFAOYSA-N

Application

Precursor commonly used in the synthesis of biologically active compounds including:
  • Multisubstituted purines for use as P2X7 antagonists in the treatment of pain
  • Heteroaryl substituted tetrahydropyrroloijquinolinone derivatives as aldosterone synthase inhibitors
  • Fluorohydroquinolineethanol as a CETP inhibitor
  • Biaryl amides with muscarinic acetylcholine receptor subtype M1 agonist activity
  • C2-aryl pyrrolobenzodiasepine antitumor agents
  • Piperazine-bisamide for obesity treatments

Other Notes

Contains varying amounts of anhydride

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service