Skip to Content
Merck
All Photos(2)

Key Documents

D43107

Sigma-Aldrich

2,5-Dibromopyridine

98%

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
$126.00

$126.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
10 G
$126.00

About This Item

Empirical Formula (Hill Notation):
C5H3Br2N
CAS Number:
Molecular Weight:
236.89
Beilstein:
109099
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$126.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request a Bulk Order

Assay

98%

form

powder

mp

92-95 °C (lit.)

SMILES string

Brc1ccc(Br)nc1

InChI

1S/C5H3Br2N/c6-4-1-2-5(7)8-3-4/h1-3H

InChI key

ZHXUWDPHUQHFOV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Minami Kato et al.
ChemSusChem, 13(9), 2379-2385 (2020-02-11)
Many types of batteries have been proposed as next-generation energy-storage systems. One candidate is a rocking-chair-type "molecular ion battery" in which a molecular ion, instead of Li+ , works as a charge carrier. Previously, we reported a viologen-type derivative as

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service