Skip to Content
Merck
All Photos(1)

Key Documents

375063

Sigma-Aldrich

1-Adamantyl isocyanate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15NO
CAS Number:
Molecular Weight:
177.24
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

97%

mp

144-146 °C (lit.)

functional group

isocyanate

SMILES string

O=C=NC12C[C@H]3C[C@H](C[C@H](C3)C1)C2

InChI

1S/C11H15NO/c13-7-12-11-4-8-1-9(5-11)3-10(2-8)6-11/h8-10H,1-6H2/t8-,9+,10-,11-

InChI key

VBHCPGFCIQDXGZ-BIBSGERRSA-N

General description

1-Adamantyl isocyanate is an isocyanate derivative.

Application

1-Adamantyl isocyanate may be used in the preparation of:
  • adamantyl-functionalized nanoparticles, via thiol-isocyanate reaction of thiol-modified nanoparticles1 4-[3-(aryl or -cycloalkyl)ureido]butyryl compound[1]
  • 4-(3-adamantan-1-yl-ureido)butyric acid, via reaction with 4-aminobutanoic acid in DMF[2]
  • 4-(3-adamantan-1-yl-ureido)butyryloxy compound[2]
  • 12-(3-adamantan-1-yl-ureido) dodecanoic acid (AUDA)[3]
  • adamantyl-phenylsulfonamide ureas[4]
  • 1-adamantan-1-yl-3-(hydroxyalkyl)ureas, via reaction with with hydroxyalkylamines[5]

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

In-Hae Kim et al.
Journal of medicinal chemistry, 47(8), 2110-2122 (2004-04-02)
The soluble epoxide hydrolase (sEH) is involved in the metabolism of endogenous chemical mediators that play an important role in blood pressure regulation and inflammation. 1,3-Disubstituted ureas are potent inhibitors of sEH that are active both in vitro and in
E Jeffrey North et al.
Bioorganic & medicinal chemistry, 21(9), 2587-2599 (2013-03-19)
Out of the prominent global ailments, tuberculosis (TB) is still one of the leading causes of death worldwide due to infectious disease. Development of new drugs that shorten the current tuberculosis treatment time and have activity against drug resistant strains
Kathrin Isenbügel et al.
Macromolecular rapid communications, 31(24), 2121-2126 (2011-05-14)
Stable nanoparticle vesicles were for the first time prepared from adamantyl- and cyclodextrin (CD)-modified silica nanoparticles forming host-guest interactions in aqueous solution. Adamantyl-functionalized nanoparticles were obtained from thiol-isocyanate reaction of thiol-modified nanoparticles with 1-adamantyl isocyanate. The CD modified silica particles
In-Hae Kim et al.
Bioorganic & medicinal chemistry, 15(1), 312-323 (2006-10-19)
The soluble epoxide hydrolase (sEH) plays an important role in the metabolism of endogenous chemical mediators involved in blood pressure regulation and vascular inflammation. 12-(3-Adamantan-1-yl-ureido)-dodecanoic acid (AUDA, 1) is a very active inhibitor of sEH both in vitro and in
In-Hae Kim et al.
Journal of medicinal chemistry, 50(21), 5217-5226 (2007-09-27)
Soluble epoxide hydrolase (sEH) is a therapeutic target for treating hypertension and inflammation. 1,3-Disubstituted ureas functionalized with an ether group are potent sEH inhibitors. However, their relatively low metabolic stability leads to poor pharmacokinetic properties. To improve their bioavailability, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service