Skip to Content
Merck
All Photos(1)

Key Documents

49546

Sigma-Aldrich

Ethyl 2-trans-4-cis-decadienoate

technical, ≥97.0% (sum of isomers, GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH=CHCH=CHCO2C2H5
CAS Number:
Molecular Weight:
196.29
Beilstein:
1724176
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Assay

≥85% 2E,4Z-isomer basis (GC)
≥97.0% (sum of isomers, GC)

refractive index

n20/D 1.486 (lit.)
n20/D 1.487

bp

70-72 °C/0.05 mmHg (lit.)

density

0.905 g/mL at 25 °C (lit.)

SMILES string

[H]\C(CCCCC)=C([H])\C([H])=C\C(=O)OCC

InChI

1S/C12H20O2/c1-3-5-6-7-8-9-10-11-12(13)14-4-2/h8-11H,3-7H2,1-2H3/b9-8-,11-10+

InChI key

OPCRGEVPIBLWAY-QNRZBPGKSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ethyl (E,Z)-2,4-decadienoate
Tsuboi S, et al.
Organic Syntheses, 66, 22-22 (1988)
Factors affecting the differential capture of male and female codling moth (Lepidoptera: Tortricidae) in traps baited with ethyl (E,Z)-2,4-decadienoate.
Knight AL and Light DM.
Environmental Entomology, 34(5), 1161-1169 (2005)
P J Landolt et al.
Journal of chemical ecology, 33(12), 2236-2244 (2007-11-10)
Codling moths are attracted to acetic acid and to ethyl-(E,Z)-2,4-decadienoate, the pear ester, when presented individually. The attraction to acetic acid is thought to be a food finding behavior, whereas the pear odorant, ethyl-(E,Z)-2,4-decadienoate, may be a host kairomone. We
Silvia Schmidt et al.
Environmental entomology, 36(5), 1025-1031 (2008-02-21)
Ethyl (E,Z)-2,4-decadienoate (pear ester) is known to be a kairomonal attractant for both male and female codling moths, Cydia pomonella (L.) (Lepidoptera: Tortricidae). Electroantennographic (EAG) studies were conducted to investigate the sensitivities of codling moth, the chestnut torticid species Cydia

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service