Skip to Content
Merck
All Photos(1)

Documents

W349801

Sigma-Aldrich

cis-3-Hexenyl 3-methylbutanoate

≥97%, FCC, FG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2CHCH2CO2CH2CH2CH=CHC2H5
CAS Number:
Molecular Weight:
184.28
FEMA Number:
3498
EC Number:
Council of Europe no.:
2344
MDL number:
UNSPSC Code:
12164502
PubChem Substance ID:
Flavis number:
09.505
NACRES:
NA.21

biological source

synthetic

Quality Level

grade

FG
Halal
Kosher

reg. compliance

EU Regulation 1334/2008 & 178/2002
FCC

Assay

≥97%

refractive index

n20/D 1.432 (lit.)

bp

98 °C/15 mmHg (lit.)

density

0.874 g/mL at 25 °C (lit.)

application(s)

flavors and fragrances

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

Organoleptic

apple; green

SMILES string

CC\C=C/CCOC(=O)CC(C)C

InChI

1S/C11H20O2/c1-4-5-6-7-8-13-11(12)9-10(2)3/h5-6,10H,4,7-9H2,1-3H3/b6-5-

InChI key

AIQLNKITFBJPFO-WAYWQWQTSA-N

Related Categories

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

140.0 °F - closed cup

Flash Point(C)

60 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Martin Heil et al.
Journal of chemical ecology, 34(5), 601-604 (2008-04-15)
Herbivore-induced volatile organic compounds (VOCs) are widely appreciated as an indirect defense mechanism since carnivorous arthropods use VOCs as cues for host localization and then attack herbivores. Another function of VOCs is plant-plant signaling. That VOCs elicit defensive responses in
Riikka Rinnan et al.
Frontiers in microbiology, 4, 224-224 (2013-08-24)
Biogenic volatile organic compounds (BVOCs) affect both atmospheric processes and ecological interactions. Our primary aim was to differentiate between BVOC emissions from above- and belowground plant parts and heath soil outside the growing season. The second aim was to assess

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service