Skip to Content
Merck
All Photos(1)

Key Documents

493031

Sigma-Aldrich

Phenacetin-ethoxy-2-13C

99 atom % 13C

Synonym(s):

N-(4-Ethoxy-2-13C-phenyl)acetamide

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$2,050.00

$2,050.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request more information

Select a Size

Change View
500 MG
$2,050.00

About This Item

Linear Formula:
CH3CONHC6H4OCH213CH3
CAS Number:
Molecular Weight:
180.21
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.12

$2,050.00

PRICE WITHOUT NATIONAL TAXES

Estimated to ship onJune 02, 2025


Request more information

isotopic purity

99 atom % 13C

mp

134-136 °C (lit.)

mass shift

M+1

SMILES string

CC(=O)Nc1ccc(OC[13CH3])cc1

InChI

1S/C10H13NO2/c1-3-13-10-6-4-9(5-7-10)11-8(2)12/h4-7H,3H2,1-2H3,(H,11,12)/i1+1

InChI key

CPJSUEIXXCENMM-OUBTZVSYSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

187.9 °F - Pensky-Martens closed cup

Flash Point(C)

86.6 °C - Pensky-Martens closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service