Skip to Content
Merck
All Photos(1)

Key Documents

490903

Sigma-Aldrich

Thiourea-13C,15N2

99 atom % 13C, 98 atom % 15N

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$3,290.00

$3,290.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
500 MG
$3,290.00

About This Item

Linear Formula:
H215N13CS15NH2
CAS Number:
Molecular Weight:
79.10
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

$3,290.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

99 atom % 13C
98 atom % 15N

Assay

99% (CP)

form

solid

mp

170-176 °C (lit.)

mass shift

M+3

SMILES string

[15NH2][13C]([15NH2])=S

InChI

1S/CH4N2S/c2-1(3)4/h(H4,2,3,4)/i1+1,2+1,3+1

InChI key

UMGDCJDMYOKAJW-VMIGTVKRSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service