Skip to Content
Merck
All Photos(1)

Key Documents

615854

Sigma-Aldrich

N-Methyl-d3-2-pyrrolidinone-d6

98 atom % D

Synonym(s):

1-Methyl-d3-2-pyrrolidinone-d6, Deuterated methylpyrrolidinone

Sign Into View Organizational & Contract Pricing

Select a Size

1 G
$617.00

$617.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
1 G
$617.00

About This Item

Empirical Formula (Hill Notation):
C5D9NO
CAS Number:
Molecular Weight:
108.19
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12

$617.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

98 atom % D

Quality Level

Assay

99% (CP)

bp

202 °C (lit.)

density

1.125 g/mL at 25 °C

mass shift

M+9

SMILES string

[2H]C([2H])([2H])N1C(=O)C([2H])([2H])C([2H])([2H])C1([2H])[2H]

InChI

1S/C5H9NO/c1-6-4-2-3-5(6)7/h2-4H2,1H3/i1D3,2D2,3D2,4D2

InChI key

SECXISVLQFMRJM-YNSOAAEFSA-N

Related Categories

General description

N-Methyl-d3-2-pyrrolidinone-d9 is isotopically labeled analogue of N-methyl-2-pyrrolidinone

Application

N-Methyl-d3-2-pyrrolidinone-d9 can be used to investigate the mechanistic study of base-catalyzed carbon-alkylation of potassium enolates with styrenes via a metal–ene reaction.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

186.8 °F - closed cup

Flash Point(C)

86 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joshua P Barham et al.
Organic & biomolecular chemistry, 18(11), 2063-2075 (2020-02-27)
Base-catalyzed, C-alkylation of potassium (K) enolates with styrenes (CAKES) has recently emerged as a highly practical and convenient method for elaboration or synthesis of pharmaceutically-relevant cores. K enolate-type precursors such as alkyl-substituted heterocycles (pyridines, pyrazines and thiophenes), ketones, imines, nitriles

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service