Skip to Content
Merck
All Photos(1)

Key Documents

490172

Sigma-Aldrich

L-Valine-15N

98 atom % 15N

Synonym(s):

(S)-α-Aminoisovaleric acid-15N, L-2-Amino-3-methylbutanoic acid-15N, 15N Labeled L-valine

Sign Into View Organizational & Contract Pricing

Select a Size

500 MG
$800.00

$800.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

Select a Size

Change View
500 MG
$800.00

About This Item

Linear Formula:
(CH3)2CHCH(15NH2)CO2H
CAS Number:
Molecular Weight:
118.14
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.12

$800.00

PRICE WITHOUT NATIONAL TAXES

Check Cart for Availability

Request more information

isotopic purity

98 atom % 15N

Quality Level

Assay

99% (CP)

form

solid

optical activity

[α]25/D +26.6°, c = 1 in 5 M HCl

technique(s)

bio NMR: suitable

mp

295-300 °C (subl.) (lit.)

mass shift

M+1

SMILES string

CC(C)[C@H]([15NH2])C(O)=O

InChI

1S/C5H11NO2/c1-3(2)4(6)5(7)8/h3-4H,6H2,1-2H3,(H,7,8)/t4-/m0/s1/i6+1

InChI key

KZSNJWFQEVHDMF-JGTYJTGKSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Nadejda Bozadjieva Kramer et al.
Cell reports, 33(2), 108239-108239 (2020-10-15)
Elevated levels of branched-chain amino acids (BCAAs) and their metabolites are strongly positively associated with obesity, insulin resistance, and type 2 diabetes. Bariatric surgery is among the best treatments for weight loss and associated morbidities. Clinical studies have reported that

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service